Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-(trifluoromethylthio)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Bromophenyl trifluoromethyl sulphide4-(Trifluoromethylthio)bromobenzene

B20874-06 B20874-03 B20874-06 B20874-03
AAB20874-03EA 39.29 USD InStock
CAAAB20874-06 CAAAB20874-03 AAB20874-03 AAB20874-06
1-Bromo-4-(trifluoromethylthio)benzene 97%
1-Bromo-4-(trifluoromethylthio)benzene


Formula: C₇H₄BrF₃S
MW: 257.07 g/mol
Boiling Pt: 199…200 °C
Density: 1.710
Flash Pt: 83 °C(181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040840
CAS Number: 333-47-1
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty