Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-4'-nitrostilbene 98%

Supplier: Thermo Scientific Chemicals
Warning

B20913-06 B20913-03 B20913-03 B20913-06
AAB20913-03EA 56.19 USD InStock
CAAAB20913-03 AAB20913-03 AAB20913-06 CAAAB20913-06
4-Hydroxy-4'-nitrostilbene 98%
4-Hydroxy-4'-nitrostilbene


Formula: C₁₄H₁₁NO₃
MW: 241.25 g/mol
Melting Pt: 205…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00017044
CAS Number: 19221-08-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty