Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-2-(difluoromethoxy)toluene 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(Difluoromethoxy)benzyl bromide o-(Difluoromethoxy)benzyl bromide

B20931-18 B20931-18 B20931-09 B20931-09
AAB20931-09EA 300.99 USD InStock
CAAAB20931-09 AAB20931-09 AAB20931-18 CAAAB20931-18
α-Bromo-2-(difluoromethoxy)toluene 97%
α-Bromo-2-(difluoromethoxy)toluene


Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Boiling Pt: 222 °C
Melting Pt: 13 °C
Density: 1.555
Flash Pt: 108 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190103
CAS Number: 85684-64-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty