Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-5-phenyl-3-furoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methyl-5-phenylfuran-3-carboxylic acid

B20988-06 B20988-06 B20988-03 B20988-03
AAB20988-03EA 107.99 USD InStock
AAB20988-03 CAAAB20988-03 CAAAB20988-06 AAB20988-06
2-Methyl-5-phenyl-3-furoic acid 97%
2-Methyl-5-phenyl-3-furoic acid


Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00221070
CAS Number: 108124-17-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty