Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactitol monohydrate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3R,4R,5R)-4-(((2S,3R,4S,5R,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol hydrate

B21016-06 B21016-03 B21016-06 B21016-03
AAB21016-03EA 61.39 USD InStock
CAAAB21016-06 AAB21016-03 AAB21016-06 CAAAB21016-03
Lactitol monohydrate 98%
Lactitol monohydrate


Formula: C₁₂H₂₄O₁₁.H₂O
MW: 362.33 g/mol (344.31 anhy)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00150767
CAS Number: 81025-04-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty