Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Fmoc-glycine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Gly-OH2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid

B21050-06 B21050-06 B21050-03 B21050-03
AAB21050-03EA 28.1 USD InStock
AAB21050-03 AAB21050-06 CAAAB21050-06 CAAAB21050-03
N-Fmoc-glycine 98%
N-Fmoc-glycine


Formula: C₁₇H₁₅NO₄
MW: 297.3 g/mol
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00037140
CAS Number: 29022-11-5
EINECS: 249-373-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty