Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salmon GlcA CHA salt, salmon-Gluc CHA salt 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 6-Chloro-3-indolyl-beta-D-glucuronic acid cyclohexylammonium salt

B21068-MF B21068-MF B21068-MA B21068-MA
AAB21068-MAEA 39.22 USD InStock
AAB21068-MA AAB21068-MF CAAAB21068-MA CAAAB21068-MF
Salmon GlcA CHA salt, salmon-Gluc CHA salt 98%
Salmon GlcA CHA salt, salmon-Gluc CHA salt


Formula: C₁₄H₁₄ClNO₇·C₆H₁₃N
MW: 442.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153940
CAS Number: 138182-20-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty