Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl yellow ≥98% indicator

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dimethyl yellowSudan Yellow 4-(Dimethylamino)azobenzene 4-(N,N-Dimethylamino)azobenzene

B21145-36 B21145-22 B21145-14 B21145-22 B21145-14 B21145-36
AAB21145-36EA 295.99 USD InStock
CAAAB21145-14 CAAAB21145-36 AAB21145-36 AAB21145-14 AAB21145-22 CAAAB21145-22
Methyl yellow ≥98% indicator
Methyl Yellow


Formula: C₁₄H₁₅N₃
MW: 225.29 g/mol
Melting Pt: 110…112 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 3143
ADR: "6.1",III
Merck Index: 12,03279
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty