Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl adipate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1,6-Hexanedioic acid dimethyl ester Adipic acid dimethyl ester

B21174-30 B21174-18 B21174-0B
AAB21174-0BEA 111.82 USD InStock
AAB21174-0B AAB21174-30 AAAB21174-18
Dimethyl adipate 99%
Dimethyl adipate


Formula: C₈H₁₄O₄
Boiling Pt: 109…110 °C/14 mm
Melting Pt: 8 °C
Density: 1,063
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008469
CAS Number: 627-93-0
EINECS: 211-020-6
Merck Index: 13,00163
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty