Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl dichlorophosphite 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethyl phosphorodichloridite

B21179-18 B21179-09 B21179-30 B21179-09 B21179-18 B21179-30
AAB21179-09EA 80.99 USD InStock
AAB21179-09 AAB21179-18 AAAB21179-30 CAAAAB21179-30 CAAAB21179-18 CAAAB21179-09
Ethyl dichlorophosphite 98%
Ethyl dichlorophosphite


Formula: C₂H₅Cl₂OP
Boiling Pt: 117…118 °C
Density: 1.284
Flash Pt: 4 °C(39 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000525
CAS Number: 1498-42-6
EINECS: 216-098-5
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty