Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Aspartic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(-)-2-Amino-1,4-butanedioic acid (R)-(-)-Aspartic acid D(-)-2-Amino-1,4-butanedioic acid H-D-Asp-OH

B21184-06 B21184-22 B21184-06 B21184-14 B21184-14 B21184-22
AAB21184-22EA 268.99 USD InStock
CAAAB21184-06 AAB21184-22 AAB21184-06 AAB21184-14 CAAAB21184-22 CAAAB21184-14
D-(-)-Aspartic acid 99%
D(-)-Aspartic acid


Formula: C₄H₇NO₄
MW: 133.1 g/mol
Melting Pt: >300 °C (dec.)
Density: 1.66
Storage Temperature: Ambient
MDL Number: MFCD00063081
CAS Number: 1783-96-6
EINECS: 217-234-6
Merck Index: 13,00845
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty