Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-MUG 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-methylumbelliferyl-beta-D-glucuronide

B21190-MD B21190-MF B21190-MD B21190-MF
AAB21190-MFEA 74.46 USD InStock
AAB21190-MF AAB21190-MD CAAAB21190-MF CAAAB21190-MD
4-MUG 98%
4-MUG


Formula: C₁₆H₁₆O₉
MW: 352.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036772
CAS Number: 6160-80-1
EINECS: 228-186-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty