Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Sorbose 98%

Supplier: Thermo Scientific Chemicals

B21208-02 B21208-77 B21208-77 B21208-02
AAB21208-77EA 95.99 USD InStock
CAAAB21208-02 CAAAB21208-77 AAB21208-77 AAB21208-02
D-(+)-Sorbose 98%
D(+)-Sorbose


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00151095
CAS Number: 3615-56-3
EINECS: 222-796-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty