Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Pentanol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DL-2-Pentanol (±)-2-Hydroxy pentane (±)-Methyl propyl carbinol (±)-Propyl methyl carbinol

B21217-0F B21217-0F B21217-AP B21217-AP
AAB21217-APEA 83.8 USD InStock
CAAAB21217-0F AAB21217-AP AAB21217-0F CAAAB21217-AP
(±)-2-Pentanol 99%
(±)-2-Pentanol


Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 114…115 °C
Melting Pt: –50 °C
Density: 0.811
Flash Pt: 33 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004579
CAS Number: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: "3",III
Merck Index: 12,07258
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/04/710504.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty