Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Turanose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one

B21224-06 B21224-06 B21224-03 B21224-03
AAB21224-06EA 217.99 USD InStock
AAB21224-06 CAAAB21224-03 CAAAB21224-06 AAB21224-03
D-(+)-Turanose 98%
D(+)-Turanose
Sucrose analog not metabolized by higher plants. Carbon source for fungi and bacteria


Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: ca 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006606
CAS Number: 547-25-1
EINECS: 208-918-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty