Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D(+)-Talose 97%

Supplier: Thermo Scientific Chemicals

B21226-77 B21226-02 B21226-77 B21226-02
AAB21226-02EA 376.99 USD InStock
CAAAB21226-77 CAAAB21226-02 AAB21226-02 AAB21226-77
α-D(+)-Talose 97%
α-D-Talose


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00064906
CAS Number: 2595-98-4
EINECS: 219-996-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty