Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Thioguanine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Amino-6-mercaptopurine6-Mercaptoguanine

B21280-06 B21280-03 B21280-03 B21280-06
AAB21280-03EA 86.29 USD InStock
CAAAB21280-03 AAB21280-03 CAAAB21280-06 AAB21280-06
6-Thioguanine 98%
6-Thioguanine


Formula: C₅H₅N₅S
MW: 167.19 g/mol
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00233553
CAS Number: 154-42-7
EINECS: 205-827-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty