Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl terephthalate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Diethyl benzene-1,4-dicarboxylate

B21401-18 B21401-09 B21401-18 B21401-09
AAB21401-09EA 39.29 USD InStock
CAAAB21401-09 CAAAB21401-18 AAB21401-09 AAB21401-18
Diethyl terephthalate 95%
Diethyl terephthalate


Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 44…46 °C
Density: 1.100
Storage Temperature: Ambient
MDL Number: MFCD00039891
CAS Number: 636-09-9
EINECS: 211-249-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty