Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl tetradecylphosphonate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Diethyl 1-tetradecylphosphonate

B21426-06 B21426-14 B21426-06 B21426-14
AAB21426-06EA 85.19 USD InStock
AAB21426-06 AAB21426-14 CAAAB21426-14 CAAAB21426-06
Diethyl tetradecylphosphonate 98%
Diethyl tetradecylphosphonate


Formula: C₁₈H₃₉O₃P
Boiling Pt: 199…201 °C/3 mm
Melting Pt: 8 °C
Density: 0.922
Storage Temperature: Ambient
MDL Number: MFCD00015597
CAS Number: 5191-09-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty