Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylcalix[8]arene 96%

Supplier: Thermo Scientific Chemicals
Warning

B21443-06 B21443-03 B21443-06 B21443-14 B21443-14 B21443-03
AAB21443-03EA 43.79 USD InStock
AAB21443-03 AAB21443-14 AAB21443-06 CAAAB21443-06 CAAAB21443-14 CAAAB21443-03
4-tert-Butylcalix[8]arene 96%
4-tert-Butylcalix[8]arene


Formula: C₈₈H₁₁₂O₈
Melting Pt: 411…412 °C
Storage Temperature: Ambient
MDL Number: MFCD00075392
CAS Number: 68971-82-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty