Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Asparagine, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-(+)-Asparagine

B21473-0E B21473-36 B21473-36 B21473-22 B21473-22 B21473-0E
AAB21473-0EEA 805.99 USD InStock
AAB21473-0E AAB21473-36 CAAAB21473-22 CAAAB21473-36 AAB21473-22 CAAAB21473-0E
L(+)-Asparagine, anhydrous 99%
L(+)-Asparagine


Formula: H₂NCOCH₂CH(NH₂)CO₂H
MW: 132.12 g/mol
Melting Pt: 234…236 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064401
CAS Number: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://us-legacy.vwr.com/stibo/web/std.lang.all/32/67/16113267.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty