Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-o-xylene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-2,3-dimethylbenzene 3-Chloro-1,2-dimethylbenzene

B21505-14 B21505-22 B21505-14 B21505-22
AAB21505-22EA 178.99 USD InStock
AAB21505-22 CAAAB21505-14 AAB21505-14 CAAAB21505-22
3-Chloro-o-xylene 97%
3-Chloro-o-xylene


Formula: C₈H₉Cl
MW: 140.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060689
CAS Number: 608-23-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty