Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Butoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals

B21506-14 B21506-22 B21506-14 B21506-22
AAB21506-14EA 76.79 USD InStock
AAB21506-14 CAAAB21506-14 AAB21506-22 CAAAB21506-22
4-Butoxybenzoic acid 98%
4-Butoxybenzoic acid


Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Melting Pt: 146…147 °C (C-Nematic)
Storage Temperature: Ambient
MDL Number: MFCD00002546
CAS Number: 1498-96-0
EINECS: 216-104-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty