Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nonafluorovaleric acid 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: IPC-PFFA-5Nonafluoropentanoic AcidPerfluoropentanoic AcidPerfluorovaleric Acid

B21567-14 B21567-14 B21567-06 B21567-06
AAB21567-14EA 263 USD InStock
AAB21567-14 CAAAB21567-06 AAB21567-06 CAAAB21567-14
Nonafluorovaleric acid 97%
Nonafluorovaleric acid


Formula: C₅HF₉O₂
MW: 264.05 g/mol
Boiling Pt: 140 °C
Density: 1.713
Storage Temperature: Ambient
MDL Number: MFCD00040211
CAS Number: 2706-90-3
EINECS: 220-300-7
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty