Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pelargonic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Nonanoic acid

B21568-AU B21568-AK B21568-AU B21568-AK
AAB21568-AUEA 65.17 USD InStock
AAB21568-AU CAAAB21568-AK AAB21568-AK CAAAB21568-AU
Pelargonic acid 97%
Pelargonic acid


Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 253…255 °C
Melting Pt: 10…12 °C
Density: 0.905
Flash Pt: 140 °C(284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004433
CAS Number: 112-05-0
EINECS: 203-931-2
ADR: 8,III
Merck Index: 12,07198
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty