Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Curcumin (from turmeric rhizome) 95% (total curcuminoid content)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

B21573-09 B21573-18 B21573-18 B21573-09 B21573-30 B21573-30
AAB21573-09EA 86.63 USD InStock
AAB21573-09 CAAAB21573-30 AAB21573-18 CAAAB21573-09 CAAAB21573-18 AAB21573-30
Curcumin (from turmeric rhizome) 95% (total curcuminoid content)
Curcumin
Natural Yellow 3. Inhibits lipoxygenase and cyclooxygenase


Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty