Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

B21658-14 B21658-06 B21658-14 B21658-03 B21658-06 B21658-03
AAB21658-06EA 46.49 USD InStock
CAAAB21658-14 CAAAB21658-03 CAAAB21658-06 AAB21658-06 AAB21658-14 AAB21658-03
2-Bromo-6-nitrotoluene 98%
2-Bromo-6-nitrotoluene


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 108…110 °C/3.3 mm
Melting Pt: 37…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009792
CAS Number: 55289-35-5
EINECS: 259-566-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty