Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Carbobenzoxy-L-proline ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Cbz-L-prolineZ-Pro-OH

B21672-18 B21672-30 B21672-18 B21672-30 B21672-06 B21672-06
AAB21672-30EA 768.99 USD InStock
AAB21672-30 AAAB21672-06 CAAAB21672-18 CAAAAB21672-06 CAAAB21672-30 AAB21672-18
N-Carbobenzoxy-L-proline ≥98%
N-Carbobenzoxy-L-proline


Formula: C₁₃H₁₅NO₄
MW: 249.27 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00003170
CAS Number: 1148-11-4
EINECS: 214-557-4
Order Now
Specification Test Results
Optical rotation -60° (c=1 in acetic acid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty