Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-5-vinylthiazole ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Vinylsulfurol

B21719-22 B21719-22 B21719-14 B21719-06 B21719-06 B21719-14
AAB21719-22EA 332.99 USD InStock
AAB21719-22 CAAAB21719-14 CAAAB21719-06 CAAAB21719-22 AAB21719-06 AAB21719-14
4-Methyl-5-vinylthiazole ≥98%
4-Methyl-5-vinylthiazole


Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 67…68 °C/11 mm
Melting Pt: –15 °C
Density: 1.095
Flash Pt: 70 °C(158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005337
CAS Number: 1759-28-0
EINECS: 217-160-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty