Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Bromobutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromobutanoic acid(±)-2-Bromobutanoic acid

B21743-0B B21743-30 B21743-0B B21743-30
AAB21743-30EA 152.99 USD InStock
CAAAB21743-0B AAB21743-30 CAAAB21743-30 AAB21743-0B
(±)-2-Bromobutyric acid 98%
(±)-2-Bromobutyric acid


Formula: H₃CCH₂BrCHCOOH
MW: 167 g/mol
Boiling Pt: 99…103 °C/10 mm
Melting Pt: –4 °C
Density: 1.567
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004216
CAS Number: 80-58-0
EINECS: 201-294-5
UN: 2922
ADR: "8",III
Merck Index: 13,01397
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty