Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Nitrophenyl)methanamine hydrochloride

B21748-06 B21748-14 B21748-03 B21748-06 B21748-14 B21748-03
AAB21748-06EA 97.89 USD InStock
AAB21748-06 CAAAB21748-06 CAAAB21748-14 CAAAB21748-03 AAB21748-14 AAB21748-03
4-Nitrobenzylamine hydrochloride 97%
4-Nitrobenzylamine hydrochloride


Formula: C₇H₉ClN₂O₂
MW: 188.62 g/mol
Melting Pt: 255…257 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012863
CAS Number: 18600-42-5
EINECS: 242-441-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty