Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-1,2-Propanediol 97%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-(+)-1,2-Dihydroxy propane(S)-(+)-1,2-Propylene glykol(S)-Propane-1,2-diol

B21773-03 B21773-06 B21773-03 B21773-06 B21773-14 B21773-14
AAB21773-06EA 176.99 USD InStock
AAB21773-06 CAAAB21773-06 CAAAB21773-03 CAAAB21773-14 AAB21773-03 AAB21773-14
(S)-(+)-1,2-Propanediol 97%
(S)-(+)-1,2-Propanediol


Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 186…188 °C
Density: 1.040
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004539
CAS Number: 4254-15-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty