Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester5-(2-Acetoxyethyl)-4-methylthiazole

B21867-06 B21867-22 B21867-22 B21867-06 B21867-14 B21867-14
AAB21867-14EA 63.49 USD InStock
AAB21867-14 CAAAB21867-06 AAB21867-06 CAAAB21867-14 CAAAB21867-22 AAB21867-22
2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98%
2-(4-Methyl-5-thiazolyl)ethyl acetate


Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
Boiling Pt: 116…118 °C/6 mm
Density: 1.147
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005338
CAS Number: 656-53-1
EINECS: 211-515-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty