Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digoxin 96%

Supplier: Thermo Scientific Chemicals
Danger

B21902-06 B21902-03 B21902-03 B21902-06
AAB21902-03EA 145.52 USD InStock
AAB21902-03 CAAAB21902-03 CAAAB21902-06 AAB21902-06
Digoxin 96%
Digoxin
A cardiac glucoside and substrate for Pgp; upregulates Pgp expression.


Formula: C₄₁H₆₄O₁₄
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003674
CAS Number: 20830-75-5
EINECS: 244-068-1
UN: 3462
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty