Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Trimethylsilyl)thiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

B21903-06 B21903-14 B21903-14 B21903-03 B21903-06 B21903-03
AAB21903-06EA 186.99 USD InStock
AAB21903-06 CAAAB21903-06 CAAAB21903-14 CAAAB21903-03 AAB21903-14 AAB21903-03
2-(Trimethylsilyl)thiazole 97%
2-(Trimethylsilyl)thiazole


Formula: C₆H₁₁NSSi
MW: 157.31 g/mol
Boiling Pt: 53…55 °C/10 mm
Density: 0.985
Flash Pt: 57 °C(135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066274
CAS Number: 79265-30-8
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty