Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodoaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

B21936-18 B21936-30 B21936-18 B21936-30
AAB21936-18EA 74.97 USD InStock
CAAAB21936-18 CAAAB21936-30 AAB21936-18 AAB21936-30
4-Iodoaniline 98%
4-Iodoaniline


Formula: C₆H₆IN
MW: 219.02 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007848
CAS Number: 540-37-4
EINECS: 208-743-4
UN: 2811
ADR: "6.1",III
Merck Index: 13,05049
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty