Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-(Diethylamino)-4-methylcoumarin 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Coumarin 17-(Diethylamino)-4-methyl-2H-chromen-2-one7-Diethylamino-4-methylcoumarin

B21938-22 B21938-36 B21938-36 B21938-22
AAB21938-22EA 68.78 USD InStock
AAB21938-22 AAB21938-36 CAAAB21938-22 CAAAB21938-36
7-(Diethylamino)-4-methylcoumarin 99%
7-(Diethylamino)-4-methylcoumarin


Formula: C₁₄H₁₇NO₂
MW: 231.30 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00006864
CAS Number: 91-44-1
EINECS: 202-068-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty