Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-D-glutamic acid 1-benzyl ester 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Benzyl N-Boc-D-glutamateBoc-D-Glu-OBzlN-(tert-Butoxycarbonyl)-D-glutamic Acid 1-Benzyl EsterN-Boc-D-glutamic Acid 1-Benzyl Ester

B21944-06 B21944-03 B21944-03 B21944-06
AAB21944-03EA 76.69 USD InStock
AAB21944-03 AAB21944-06 CAAAB21944-06 CAAAB21944-03
N-Boc-D-glutamic acid 1-benzyl ester 99%
N-Boc-D-glutamic acid 1-benzyl ester


Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038266
CAS Number: 34404-30-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty