Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Octynoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Oct-2-ynoic acid

B21957-22 B21957-06 B21957-06 B21957-22
AAB21957-22EA 2052.99 USD InStock
CAAAB21957-22 AAB21957-22 AAB21957-06 CAAAB21957-06
2-Octynoic acid 98%
2-Octynoic acid


Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 148…149 °C/19 mm
Melting Pt: 5 °C
Density: 0.961
Flash Pt: 101 °C(214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004364
CAS Number: 5663-96-7
EINECS: 227-121-3
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty