Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Methyl-2,3-dichloropropionate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 2,3-dichloropropanoate

B21993-22 B21993-36 B21993-14 B21993-14 B21993-22 B21993-36
AAB21993-36EA 462.99 USD InStock
CAAAB21993-22 AAB21993-36 AAB21993-14 CAAAB21993-14 AAB21993-22 CAAAB21993-36
(±)-Methyl-2,3-dichloropropionate 98%
(±)-Methyl 2,3-dichloropropionate


Formula: C₄H₆Cl₂O₂
MW: 157 g/mol
Boiling Pt: 91…92 °C/50 mm
Density: 1.326
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000944
CAS Number: 3674-09-7
EINECS: 222-940-2
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty