Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,4,6-Tris(trifluoromethyl)-s-triazine

B22001-06 B22001-06 B22001-03
AAB22001-03EA 38.29 USD InStock
CAAAB22001-06 AAB22001-03 AAB22001-06
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine 98%
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine


Formula: C₆F₉N₃
MW: 285.07 g/mol
Boiling Pt: 95…96 °C
Density: 1.596
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00042436
CAS Number: 368-66-1
EINECS: 206-709-3
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty