Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) hydroxide 97%

Supplier: Thermo Scientific Chemicals
Danger

B22045-18 B22045-30
AAB22045-30EA 85.83 USD InStock
AAB22045-30 AAB22045-18
Cobalt(II) hydroxide 97%
Cobalt (II) hydroxide


Formula: H₂CoO₂
MW: 92.95 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016015
CAS Number: 21041-93-0
EINECS: 244-166-4
UN: 3077
ADR: "9",III
Merck Index: 13,02467
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty