Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methylenanthic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-2-Methylheptanoic acid

B22050-06 B22050-06 B22050-14 B22050-14 B22050-22 B22050-22
AAB22050-06EA 36.69 USD InStock
CAAAB22050-06 AAB22050-06 CAAAB22050-14 CAAAB22050-22 AAB22050-22 AAB22050-14
(±)-2-Methylenanthic acid 98%
(±)-2-Methylenanthic acid


Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 139…141 °C/30 mm
Density: 0.906
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142944
CAS Number: 1188-02-9
EINECS: 214-704-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty