Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Benzyloxy)-3,5-dimethylbenzaldehyde 95%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Benzyloxy-3,5-dimethylbenzaldehyde

B22067-03 B22067-06 B22067-03 B22067-06
AAB22067-03EA 44.69 USD InStock
AAB22067-03 CAAAB22067-06 CAAAB22067-03 AAB22067-06
4-(Benzyloxy)-3,5-dimethylbenzaldehyde 95%
4-(Benzyloxy)-3,5-dimethylbenzaldehyd


Formula: C₁₆H₁₆O₂
MW: 240.30 g/mol
Boiling Pt: 162…163 °C/1 mm
Density: 1.10
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00800683
CAS Number: 144896-51-5
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty