Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Trifluoromethyl)vinyl acetate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,3,3-Trifluoroprop-1-en-2-yl acetate

B22133-30 B22133-18 B22133-30 B22133-18
AAB22133-18EA 472.99 USD InStock
CAAAB22133-18 AAB22133-18 AAB22133-30 CAAAB22133-30
1-(Trifluoromethyl)vinyl acetate 97%
1-(Trifluoromethyl)vinyl acetate


Formula: C₅H₅F₃O₂
MW: 154.09
Boiling Pt: 86…88 °C
Density: 1.200
Flash Pt: 2 °C(35 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040844
CAS Number: 2247-91-8
EINECS: 218-838-2
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty