Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-3-(2-nitrophenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-Amino-3-(2-nitrophenyl)propanoic acid

B22176-14 B22176-14 B22176-22 B22176-06 B22176-06 B22176-22
AAB22176-22EA 1115.99 USD InStock
CAAAB22176-22 AAB22176-22 CAAAB22176-14 CAAAB22176-06 AAB22176-14 AAB22176-06
3-Amino-3-(2-nitrophenyl)propionic acid 98%
3-Amino-3-(2-nitrophenyl)propionic acid


Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00090356
CAS Number: 5678-48-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty