Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Hydroxymethyl)benzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: HMBA 4-Carboxybenzylalcohol

B22183-14 B22183-14 B22183-03 B22183-06 B22183-03 B22183-06
AAB22183-06EA 198.99 USD InStock
AAB22183-06 CAAAB22183-06 CAAAB22183-14 CAAAB22183-03 AAB22183-14 AAB22183-03
4-(Hydroxymethyl)benzoic acid 98+%
4-(Hydroxymethyl)benzoic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00017598
CAS Number: 3006-96-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty