Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Melezitose hydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol xhydrate

B22209-18 B22209-09 B22209-18 B22209-09
AAB22209-09EA 60.47 USD InStock
AAB22209-09 AAB22209-18 CAAAB22209-18 CAAAB22209-09
D-(+)-Melezitose hydrate 99%
D-(+)-Melezitose hydrate


Formula: C₁₈H₃₂O₁₆
MW: 522,45 g/mol
Melting Pt: ca 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149448
CAS Number: 207511-10-2
EINECS: 209-894-9
Order Now
Specification Test Results
Optical rotation +88° (c=4 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty