Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylcaprylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Ethyloctanoic acid

B22220-88 B22220-09 B22220-88 B22220-09
AAB22220-88EA 43.69 USD InStock
CAAAB22220-09 AAB22220-88 CAAAB22220-88 AAB22220-09
4-Ethylcaprylic acid 97%
4-Ethylcaprylic acid


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 142…143 °C/10 mm
Density: 0.908
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00506494
CAS Number: 16493-80-4
EINECS: 240-560-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty