Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2,6-xylidine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Chloro-2,6-dimethylaniline

B22232-09 B22232-30 B22232-18 B22232-30 B22232-18 B22232-09
AAB22232-09EA 62.69 USD InStock
AAB22232-09 CAAAB22232-30 AAB22232-18 CAAAB22232-09 CAAAB22232-18 AAB22232-30
3-Chloro-2,6-xylidine 99%
3-Chloro-2,6-xylidine


Formula: C₈H₁₀ClN
Boiling Pt: 136…138 °C/17 mm
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD02683086
CAS Number: 26829-77-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty